Order Entry
Belgium
ContactUsLinkComponent
MMP-9/MMP-13 inhibitor I ≥95% (by HPLC), Millipore®
MMP-9/MMP-13 inhibitor I ≥95% (by HPLC), Millipore®
Catalog # 444252-1
CAS Number:  
MMP-9/MMP-13 inhibitor I ≥95% (by HPLC), Millipore®
Catalog # 444252-1
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥95%

Specification Test Results

About this item

 :
 :459.55 g/mol
 :Packaged under inert gas.
Storage Temperature:Freezer
CAS Number:204140-01-2
Formula:C₂₅H₂₅N₃O₆S
MDL Number:MFCD03453559