Water, super gradient grade suitable for UPLC/UHPLC instruments
Catalog # APPC221074.1612
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- Non-volatile Matter:0.0001%
- Specific Conductance (25 °C):1.0×10⁻⁶ ohm⁻¹ cm⁻¹
- Suitability for Gradient According to ACS:Passes Test
- Gradient at 210 nm:5 mUA
- Gradient at 254 nm:0.5 mUA
- Fluorescence at 254 nm (as Quinine):1 ppb
- Fluorescence at 365 nm (as Quinine):0.5 ppb
- Transmittance at 200 nm (UV Spectrum, 1 cm Cell):≥98%
- Transmittance at 210 nm (UV Spectrum, 1 cm Cell):≥98%
- Transmittance at 254 nm (UV Spectrum, 1 cm Cell):≥99%
- Transmittance at 300 - 450 nm (UV Spectrum, 1 cm Cell):≥99%
Specification Test Results
About this item
:
:18.02 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1.000 g/cm³
:Packaged under nitrogen.
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332