ContactUsLinkComponent
Eyepiece for digital microscope
Eyepiece for digital microscope
Catalog # 632-9043
Supplier:  MOTIC
Eyepiece for digital microscope
Catalog # 632-9043
Supplier:  MOTIC
Specifications
  • Magnification:
    10× (14 mm)
  • Description:
    Widefield eyepiece
  • Pk:
    1