Orders LinkContactUsLinkComponent
Density hydrometers, S50
Density hydrometers, S50
Supplier:   Amarell
Density hydrometers, S50
Supplier:  Amarell
Certificates
Density hydrometers, S50
28 Options Available Below
Product Details & Documents
Hydrometers in a wide range of measuring scales.
  • Shot weighted body
  • Paper scale with colour code and type designation
Suitable for government verification.
Certifications: DIN 12791
Packaging: In a transparent plastic case.
Product Family Options
Product Information
  • Model
    Range
    Division
    Length
    Availability
    Price
  • AMARH811100
    Density hydrometer
    Density hydrometer
    S50
    0,60 - 0,65 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,60 €
    64,60 €
    1 * 1 ST
       64,60 €
  • AMARH811102
    Density hydrometer
    Density hydrometer
    S50
    0,65 - 0,70 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,60 €
    64,60 €
    1 * 1 ST
       64,60 €
  • AMARH811104
    Density hydrometer
    Density hydrometer
    S50
    0,70 - 0,75 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,60 €
    64,60 €
    1 * 1 ST
       64,60 €
  • AMARH811106
    Density hydrometer
    Density hydrometer
    S50
    0,75 - 0,80 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,60 €
    64,60 €
    1 * 1 ST
       64,60 €
  • AMARH811108
    Density hydrometer
    Density hydrometer
    S50
    0,80 - 0,85 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,60 €
    64,60 €
    1 * 1 ST
       64,60 €
  • AMARH811110
    Density hydrometer
    Density hydrometer
    S50
    0,85 - 0,90 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,60 €
    64,60 €
    1 * 1 ST
       64,60 €
  • AMARH811112
    Density hydrometer
    Density hydrometer
    S50
    0,90 - 0,95 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,60 €
    64,60 €
    1 * 1 ST
       64,60 €
  • AMARH811114
    Density hydrometer
    Density hydrometer
    S50
    0,95 - 1,00 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,60 €
    64,60 €
    1 * 1 ST
       64,60 €
  • AMARH811116
    Density hydrometer
    Density hydrometer
    S50
    1,00 - 1,05 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,30 €
    70,30 €
    1 * 1 ST
       70,30 €
  • AMARH811118
    Density hydrometer
    Density hydrometer
    S50
    1,05 - 1,10 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,30 €
    70,30 €
    1 * 1 ST
       70,30 €
  • AMARH811120
    Density hydrometer
    Density hydrometer
    S50
    1,10 - 1,15 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,30 €
    70,30 €
    1 * 1 ST
       70,30 €
  • AMARH811122
    Density hydrometer
    Density hydrometer
    S50
    1,15 - 1,20 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,30 €
    70,30 €
    1 * 1 ST
       70,30 €
  • AMARH811124
    Density hydrometer
    Density hydrometer
    S50
    1,20 - 1,25 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,30 €
    70,30 €
    1 * 1 ST
       70,30 €
  • AMARH811126
    Density hydrometer
    Density hydrometer
    S50
    1,25 - 1,30 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,30 €
    70,30 €
    1 * 1 ST
       70,30 €
  • AMARH811128
    Density hydrometer
    Density hydrometer
    S50
    1,30 - 1,35 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,10 €
    83,10 €
    1 * 1 ST
       83,10 €
  • AMARH811130
    Density hydrometer
    Density hydrometer
    S50
    1,35 - 1,40 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,10 €
    83,10 €
    1 * 1 ST
       83,10 €
  • AMARH811132
    Density hydrometer
    Density hydrometer
    S50
    1,40 - 1,45 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,10 €
    83,10 €
    1 * 1 ST
       83,10 €
  • AMARH811134
    Density hydrometer
    Density hydrometer
    S50
    1,45 - 1,50 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,10 €
    83,10 €
    1 * 1 ST
       83,10 €
  • AMARH811136
    Density hydrometer
    Density hydrometer
    S50
    1,50 - 1,55 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,10 €
    83,10 €
    1 * 1 ST
       83,10 €
  • AMARH811138
    Density hydrometer
    Density hydrometer
    S50
    1,55 - 1,60 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,10 €
    83,10 €
    1 * 1 ST
       83,10 €
  • AMARH811140
    Density hydrometer
    Density hydrometer
    S50
    1,60 - 1,65 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,90 €
    94,90 €
    1 * 1 ST
       94,90 €
  • AMARH811142
    Density hydrometer
    Density hydrometer
    S50
    1,65 - 1,70 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,90 €
    94,90 €
    1 * 1 ST
       94,90 €
  • AMARH811144
    Density hydrometer
    Density hydrometer
    S50
    1,70 - 1,75 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,90 €
    94,90 €
    1 * 1 ST
       94,90 €
  • AMARH811146
    Density hydrometer
    Density hydrometer
    S50
    1,75 - 1,80 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,90 €
    94,90 €
    1 * 1 ST
       94,90 €
  • AMARH811148
    Density hydrometer
    Density hydrometer
    S50
    1,80 - 1,85 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,90 €
    94,90 €
    1 * 1 ST
       94,90 €
  • AMARH811150
    Density hydrometer
    Density hydrometer
    S50
    1,85 - 1,90 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,90 €
    94,90 €
    1 * 1 ST
       94,90 €
  • AMARH811152
    Density hydrometer
    Density hydrometer
    S50
    1,90 - 1,95 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,90 €
    94,90 €
    1 * 1 ST
       94,90 €
  • AMARH811154
    Density hydrometer
    Density hydrometer
    S50
    1,95 - 2,00 g/cm³
    0,002 g/cm³
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,90 €
    94,90 €
    1 * 1 ST
       94,90 €