Order Entry
Belgium
ContactUsLinkComponent
Avantor® Alltima HP, HPLC columns, HILIC, 3 µm
Avantor® Alltima HP, HPLC columns, HILIC, 3 µm
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Avantor® Alltima HP, HPLC columns, HILIC, 3 µm
Supplier:  Avantor
   Sustainable Options Available

Specifications

  • Catalog No:
  • HICH86461
  • HICH86463
  • HICH86462
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Surface area:
  • 230 m²/g
  • 230 m²/g
  • 230 m²/g
  • USP code:
  • L3
  • L3
  • L3
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)

Specifications

Frequently Bought Together