Double bosshead, epoxy coated
Double bosshead with M10 brass T-bar bolt, Malleable iron, 30 mm
JUCHVL04822
Double bossheads made of malleable iron, epoxy coated.
: DIN 12895
- Description:Double bosshead with M10 brass T-bar bolt
- Material:Malleable iron
- For rod Ø:30 mm