Order Entry
Belgium
Orders LinkContactUsLinkComponent
Level indicator

Level indicator

Level indicator with volume scale, 200 L barrels
Catalog # MSGA13299
Supplier:   asecos
Level indicator
Catalog # MSGA13299
Supplier:  asecos
Level indicator
Level indicator

Level indicator with volume scale, 200 L barrels

Price
66,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

This level indicator is suitable for all standard 200 l barrels and non-corrosive liquids.

Specifications
  • For:
    200 L barrels
  • Description:
    Level indicator with volume scale
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.