Orders LinkContactUsLinkComponent
Pipette controller, MACROMAN™
Pipette controller, MACROMAN™
Pipette controller, MACROMAN™, 1 - 100 ml
Catalog # GILSF110120
Supplier:   GILSON
Pipette controller, MACROMAN™
Catalog # GILSF110120
Supplier:  GILSON
Pipette controller, MACROMAN™
Pipette controller, MACROMAN™
Pipette controller, MACROMAN™, 1 - 100 ml
Price
439,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Pipette controller for all plastic and glass pipettes from 1 to 100 ml.
  • Ergonomic handle featuring wide triggers for maximum comfort and stress-free pipetting
  • Lithium-Ion battery enables a longer, vibration-free use with reduced charging time
  • Increase precision by choosing your flow rate mode from slow drops to rapid dispensing
Specifications
  • Capacity:
    1 - 100 ml
  • Description:
    Pipette controller, MACROMAN™
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Filter, 0,22 µm, standard (set of 10)
    Filter, 0,22 µm, standard (set of 10)
    GILSF107204
    Log in for availability
    1 * 10 ST - 110,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette adaptor
    Pipette adaptor
    GILSF107211
    Log in for availability
    1 * 1 ST - 33,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Lithium-ion battery
    Lithium-ion battery
    GILSF107208
    Log in for availability
    1 * 1 ST - 124,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Power supply
    Power supply
    GILSF107203
    Log in for availability
    1 * 1 ST - 141,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Wall mount
    Wall mount
    GILSF107201
    Log in for availability
    1 * 1 ST - 46,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter, 0,45 µm, sterile (set of 10)
    Filter, 0,45 µm, sterile (set of 10)
    GILSF107207
    Log in for availability
    1 * 10 ST - 184,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Adjustable side stand
    Adjustable side stand
    GILSF107210
    Log in for availability
    1 * 1 ST - 27,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Magnetic wall mount
    Magnetic wall mount
    GILSF107202
    Log in for availability
    1 * 1 ST - 71,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter, 0,22 µm, sterile (set of 10)
    Filter, 0,22 µm, sterile (set of 10)
    GILSF107205
    Log in for availability
    1 * 10 ST - 184,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Nosepiece
    Nosepiece
    GILSF107209
    Log in for availability
    1 * 1 ST - 73,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Charging desk stand
    Charging desk stand
    GILSF107200
    Log in for availability
    1 * 1 ST - 147,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter, 0,45 µm, standard (set of 10)
    Filter, 0,45 µm, standard (set of 10)
    GILSF107206
    Log in for availability
    1 * 10 ST - 110,90 €