ContactUsLinkComponent
Gas syringes, series A-2, Pressure-Lok®
Gas syringes, series A-2, Pressure-Lok®
Supplier:  VICI
Certificates
Gas syringes, series A-2, Pressure-Lok®
Supplier:  VICI
Specifications
  • Catalog No:
  • VICIPS-050025
  • VICIPS-050034
  • VICIPS-050035
  • VICIPS-050032
  • VICIPS-050031
  • Series:
  • A-2
  • A-2
  • A-2
  • A-2
  • A-2
  • Max. pressure:
  • 250 psi
  • 250 psi
  • 250 psi
  • 250 psi
  • 250 psi
  • Needle length:
  • 0,028×0,005×2"
  • 0,029×0,012×2"
  • 0,029×0,012×2"
  • 0,029×0,012×2"
  • 0,029×0,012×2"