Orders LinkContactUsLinkComponent
Bloom test vessel
Bloom test vessel
Sample bottles (Duran) for Gelatine bloom testing
Catalog # 646-0035
Supplier:   DWK Life Sciences
Bloom test vessel
Catalog # 646-0035
Supplier:  DWK Life Sciences
Bloom test vessel
Bloom test vessel
Sample bottles (Duran) for Gelatine bloom testing
Price
94,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Manufactured according to DIN ISO 9665.
Specifications
  • Description:
    Sample bottles (Duran) for Gelatine bloom testing