Orders LinkContactUsLinkComponent
Disposable cuvettes
Disposable cuvettes
Supplier:   BIOCHROM
Disposable cuvettes
Supplier:  BIOCHROM
Certificates
Disposable cuvettes
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Type
    Volume
    Path length
    Window material
    Availability
    Price
  • SCLI80-3000-81
    Disposable cuvettes
    Disposable cuvettes
    Ultra micro
    70 µl
    10 mm
    UV plastic
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • SCLI80-3000-77
    Disposable cuvettes
    Disposable cuvettes
    Semi-micro
    800 µl
    10 mm
    UV plastic
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 131,00 €
    131,00 €
    1 * 100 ST
       131,00 €
  • SCLI80-2004-53
    Disposable cuvettes
    Disposable cuvettes
    Disposable
    2,5 ml
    10 mm
    PMMA
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST