DNA transfection reagent, PolyMag Neo™
PolyMag Neo™ transfection reagent is specifically designed to achieve high transfection efficiency combined with superior transgene expression level. It is composed of magnetic nanoparticles coated with cationic molecules. It is the ideal transfection reagent for a wide variety of cells.
- High transgene expression
- High transfection efficiency
- Multipurpose (various types of nucleic acids)
- Universal (primary cells and cell lines)
- Simple, ready to use and rapid
- Non toxic
- Compatible with and without serum-containing culture media
Store at +4 °C.
: This reagent needs to be used with a magnetic plate.
: Shipped at room temperature.
- Environmentally Preferable:
Frequently Bought Together