Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lids
Microplate Lids
Lid for 96-well microplates
Lid for 96-well microplates
Lid for 96-well microplates
Catalog # BOET05.041.0100
Supplier:
BOETTGER PAUL
Expand image
Lid for 96-well microplates
Catalog # BOET05.041.0100
Supplier:
BOETTGER PAUL
Specifications
Colour:
Clear
Description:
Lid
Seq:
04
Pk:
100
Specifications
About this item
Product Details
Product Details