



Hydrogen peroxide solution 30%, AR® ACS, Macron Fine Chemicals™
Catalog # 5240-20
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:PE drum
- Pk:20 L
- Seq:5302
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:29.0 - 32.0%
- For Laboratory,Research,or Manufacturing Use:≤0.0005%
- ACS - Assay (H₂O₂):29.0 - 32.0 %
- ACS - Ammonium (NH₄):≤0.0005 %
- Chloride (Cl):≤2 ppm
- ACS - Color (APHA):≤10
- ACS - Heavy Metals (as Pb):≤0.0001 %
- ACS - Iron (Fe):≤0.00005 %
- ACS - Nitrate (NO₃):≤0.0002 %
- ACS - Phosphate (PO₄):≤0.0002 %
- ACS - Residue after Evaporation:≤0.002 %
- ACS - Sulfate (SO₄):≤0.0005 %
- ACS - Titrable Acid (meq/g):≤0.0006
Specification Test Results
About this item
:
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:<0 °C
Density:1.11 g/cm³ (20 °C)
ADR:5.1,II
:34.01 g/mol
Storage Temperature:Ambient
EINECS:231-765-0
UN:2014
:GHS03
:GHS05
:GHS06
:GHS08
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333