Order Entry
Belgium
ContactUsLinkComponent
Sodium thiosulphate pentahydrate, BAKER ANALYZED®, J.T.Baker®
Sodium thiosulphate pentahydrate, BAKER ANALYZED®, J.T.Baker®
Catalog # 0319.1000
Supplier:  Avantor
CAS Number:  
Sodium thiosulphate pentahydrate, BAKER ANALYZED®, J.T.Baker®
Catalog # 0319.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    99.5 - 101.0 %
  • Insoluble Matter:
    max. 0.005 %
  • Nitrogen Compounds (as N):
    max. 0.002 %
  • pH of 5% Solution at 25°C:
    6.0 - 8.4
  • Sulfate and Sulfite (as SO4):
    max. 0.1 %
  • Sulfide (S):
    max. 1 ppm

Specification Test Results

About this item

 :
 :248.19 g/mol
Merck Index:12,08844
Melting Pt:48 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:10102-17-7
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186