Order Entry
Belgium
Orders LinkContactUsLinkComponent
Insert tubes for scintillation vials

Insert tubes for scintillation vials

Insert tubes with caps for scintillation vials, 5,00 ml, 14×56 mm, Yes
Catalog # RATI5810110
Supplier:   RATIOLAB
Insert tubes for scintillation vials
Catalog # RATI5810110
Supplier:  RATIOLAB
Insert tubes for scintillation vials
Insert tubes for scintillation vials

Insert tubes with caps for scintillation vials, 5,00 ml, 14×56 mm, Yes

Price
205,00 €
Log in for availability
1 * 2.000 ST
Product Details & Documents
Specifications
  • Cap included:
    Yes
  • Ø×H:
    14×56 mm
  • Capacity:
    5,00 ml
  • Description:
    Insert tubes with caps for scintillation vials
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.