
Taurine ≥98.5%, Ultrapure for molecular biology
Catalog # J75824.A1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (unspecified) Absorbance (260 nm, 0.5 M, 1 cm):≤0.2
- Assay (unspecified) Absorbance (280 nm, 0.5 M, 1 cm):≤0.2
- Assay (unspecified) Ammonia (NH₃):≤0.02%
- Assay (unspecified) Use Test:Functionally Tested for Electrophoresis
- Assay (unspecified):≥98.5%
- Chloride Content:≤0.01%
- Identification (FTIR):Conforms
- Residue on Ignition:≤0.1%
- Impurity Content Sulphate (SO₄):≤0.01%
- pH (5%, H₂O):4.0 to 7.0
- Solution Test (0.5 M, H₂O):Clear and Colourless
- Arsenic (As):≤2 ppm
- Heavy Metals:≤10 ppm
Specification Test Results
About this item
:
:125.15 g/mol
Merck Index:14,09074
Melting Pt:>300 °C
EINECS:203-483-8
:GHS07
Storage Temperature:Ambient
CAS Number:107-35-7
Formula:C₂H₇NO₃S
MDL Number:MFCD00008197