Order Entry
Belgium
ContactUsLinkComponent
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Flammable
Health
Toxic
Environment
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Toxic
Environment
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on anhydrous):
    ≥ 99 %
  • Acidity:
    ≤ 0.0002 meq/g
  • Alkalinity:
    ≤ 0.0002 meq/g
  • Evaporation residue:
    ≤ 1 ppm
  • Water:
    ≤ 50 ppm
  • Ag (Silver):
    ≤ 10 ppb
  • Al (Aluminium):
    ≤ 25 ppb
  • Ca (Calcium):
    ≤ 25 ppb
  • Cu (Copper):
    ≤ 10 ppb
  • Fe (Iron):
    ≤ 10 ppb
  • K (Potassium):
    ≤ 10 ppb
  • Mg (Magnesium):
    ≤ 10 ppb
  • Mn (Manganese):
    ≤ 10 ppb
  • Na (Sodium):
    ≤ 50 ppb
  • Ni (Nickel):
    ≤ 10 ppb
  • Pb (Lead):
    ≤ 10 ppb
  • Zn (Zinc):
    ≤ 10 ppb
  • Transmittance (190 nm):
    ≥ 10 %
  • Transmittance (205 nm):
    ≥ 50 %
  • Transmittance (220 nm):
    ≥ 80 %
  • Transmittance (235 nm):
    ≥ 95 %
  • Transmittance (255 nm):
    ≥ 99 %
  • Packaged under nitrogen:
    Confirmed

Specification Test Results

Frequently Bought Together