Order Entry
Belgium
ContactUsLinkComponent
n-Heptane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Flammable
Health
Irritant
Environment
n-Heptane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
Environment
n-Heptane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    ≥ 99 %
  • Acidity:
    ≤ 0.0002 meq/g
  • Alkalinity:
    ≤ 0.0002 meq/g
  • Evaporation residue:
    ≤ 1 ppm
  • Water:
    ≤ 50 ppm
  • Ag (Silver):
    ≤ 10 ppb
  • Al (Aluminium):
    ≤ 25 ppb
  • Ca (Calcium):
    ≤ 25 ppb
  • Cu (Copper):
    ≤ 10 ppb
  • Fe (Iron):
    ≤ 10 ppb
  • K (Potassium):
    ≤ 10 ppb
  • Mg (Magnesium):
    ≤ 10 ppb
  • Mn (Manganese):
    ≤ 10 ppb
  • Na (Sodium):
    ≤ 50 ppb
  • Ni (Nickel):
    ≤ 10 ppb
  • Pb (Lead):
    ≤ 10 ppb
  • Zn (Zinc):
    ≤ 10 ppb
  • Transmittance (195 nm):
    ≥ 10 %
  • Transmittance (210 nm):
    ≥ 50 %
  • Transmittance (220 nm):
    ≥ 80 %
  • Transmittance (245 nm):
    ≥ 95 %
  • Transmittance (290 nm):
    ≥ 99 %

Specification Test Results

Frequently Bought Together