L(+)-Glutamine ≥98.5%, Ultrapure
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
- CAS Number:56-85-9
- Formula:C₅H₁₀N₂O₃
- MDL Number:MFCD00008044
- Molecular Weight:146.15 g/mol
- Merck Index:14,04471
- Boiling Pt:355 °C (1013 hPa)
- Melting Pt:185…186 °C
- Density:1.5326 g/cm³ (130 °C)
- Storage Temperature:Ambient
- EINECS:200-292-1
4 Options Available Below
Product Details & Documents
Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
- CAS Number:56-85-9
- Formula:C₅H₁₀N₂O₃
- MDL Number:MFCD00008044
- Molecular Weight:146.15 g/mol
- Merck Index:14,04471
- Boiling Pt:355 °C (1013 hPa)
- Melting Pt:185…186 °C
- Density:1.5326 g/cm³ (130 °C)
- Storage Temperature:Ambient
- EINECS:200-292-1
Specifications for Product Family
Specification Test Results
- Assay (unspecified):≥98.5%
- Chloride Content:≤0.021%
- Identification (FTIR):Conforms
- Residue on ignition:≤0.1%
- Impurity content, Sulphate (SO₄):≤0.028%
- Loss on Drying:≤0.2%
- Optical Rotation:6.3° - 7.3° (c=4 in H₂O)
- Trace Metal, Arsenic (As):≤3 ppm
- Trace Metal, Heavy Metals (Pb):≤10 ppm
- Trace Metal, Iron (Fe):≤10 ppm
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



