Order Entry
Belgium
ContactUsLinkComponent
L(+)-Histidine monohydrochloride monohydrate 98+%
L(+)-Histidine monohydrochloride monohydrate 98+%
Catalog # J17080.A1
CAS Number:  
undefined
L(+)-Histidine monohydrochloride monohydrate 98+%
Catalog # J17080.A1
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (unspecified) - Ammonium (NH₄):
    ≤0.02%
  • Assay (unspecified):
    ≥98.0%
  • Identification (FTIR):
    Conforms
  • Residue on Ignition:
    ≤0.10%
  • Impurity Content Sulphate (SO₄):
    ≤0.03%
  • Loss on Drying:
    ≤0.3%
  • Optical Rotation (c= in 6 N HCl):
    +9.2 to 10.6°
  • Thin Layer Chromatography:
    Homogeneous
  • Arsenic (As₂O₃):
    ≤1 ppm
  • Iron (Fe):
    ≤30 ppm

Specification Test Results

About this item

 :
 :209.63 g/mol
Merck Index:14,04720
Melting Pt:240 °C (dec.)
Density:1.485 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-438-9
CAS Number:5934-29-2
Formula:C₆H₉N₃O₂·HCl·H₂O
MDL Number:MFCD00151027