ContactUsLinkComponent
SPE columns, CHROMABOND® HILIC
undefined
SPE columns, CHROMABOND® HILIC
  • Catalog No:
  • MANA730594
  • MANA730593
  • Carbon load:
  • 7%
  • 7%
  • pH range:
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 500 m²/g
  • 500 m²/g
Frequently Bought Together