Orders LinkContactUsLinkComponent
Magnetic stand
Magnetic stand
Catalog # GENO786-888
Supplier:   G-Biosciences
Magnetic stand
Catalog # GENO786-888
Supplier:  G-Biosciences
Magnetic stand
Magnetic stand
Price
353,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Magnetic stand is designed for the efficent pull down of magnetic beads in 1,5 and 2 ml micro centrifgue tubes.
  • Suitable for 8×1,5 to 2 ml tubes
  • Pulls beads to tube wall to allow removal of all supernatant
The powerful magnet pulls beads to the side of the tube, allowing for the removal of all the liquid supernatant with no contamination with beads. It can also be used for immunoprecipitations and protein purification analysis.