

Dimidium bromidedisulphite blue mixed indicator, for volumetric analysis
Catalog # APPC286330.1612
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Dimidium Bromide:80 mg
- Disulphine Blue:40 mg
- Sulphuric Acid 96%:36 ml
- Methanol:100 ml
- Water (s.q.m):1000 ml
- Identity:Passes Test
- Suitability as Indicator:Passes Test
Specification Test Results
About this item
:
Density:1.023 g/cm³
Storage Temperature:Ambient
ADR:8,III
:GHS07
:GHS08