
Sodium hydroxide ∼ 0.15 mol/l with Brij® 35 for nitrate determination
Catalog # KRAF15524.5020
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:40 g/mol
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548