ContactUsLinkComponent
Septa for Precision NMR tubes, SP Wilmad-LabGlass
Septa for Precision NMR tubes, SP Wilmad-LabGlass
Catalog # WILMWG-3891-100
Supplier:  SP Industries
Septa for Precision NMR tubes, SP Wilmad-LabGlass
Catalog # WILMWG-3891-100
Supplier:  SP Industries
Specifications
  • Description:
    Septa for tubes with 5 mm OD
  • Seq:
    01
  • Pk:
    100
Specifications
About this item
Natural rubber septa suitable for 5 mm OD thin wall Precision NMR tubes.
  • Designed to maintain excellent glass to rubber contact
  • Suitable for air-sensitive samples, catalytic studies and titration experiments
Colour: white
These septa are manufactured under 'white room' conditions with certified raw materials for consistency.