ContactUsLinkComponent
Sartorius wedge for windows
Sartorius wedge for windows
Catalog # SARBYSW02
Supplier:  Sartorius Balances
Sartorius wedge for windows
Catalog # SARBYSW02
Supplier:  Sartorius Balances
Specifications
  • Description:
    Sartorius Wedge for Windows
  • Pk:
    1
  • For:
    SART
Specifications
About this item
Sartorius wedge is a software package designed to capture data from aartorius devices into windows applications, specifically microsoft excel.
  • Eliminate manual data entry errors
  • Log data from multiple devices simultaneously
  • Capture real-time data to any Windows application
  • Parse and filter the data to match your requirements
  • Accurate, reliable, and compliant
Sartorius wedge includes two of TALtech's data capture programs: WinWedge and TCPWedge. WinWedge connects to most serial or USB devices and TCPWedge connects to most networked devices.