Sheep Monoclonal IgG
BSENS-1754-500
- Antibody type:Primary
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Format:Purified IgG
- Form:Lyophilised
- Shipping temperature:Ambient
- Purification:Protein G purified
- Pk:500 µG
Sheep serum from non-immunised animals was purified by protein G affinity chromatography. This sheep IgG fraction can be used as negative control antibody for various immunological techniques where sheep primary antibodies are used.
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: