ContactUsLinkComponent
Wooden applicator
Wooden applicator
Catalog # HERE1032018
Supplier:  HERENZ HEINZ
undefined
Wooden applicator
Catalog # HERE1032018
Supplier:  HERENZ HEINZ
Specifications
  • Ø:
    4 mm
  • Description:
    Double swabs
  • Length:
    12 mm
  • Shaft/head:
    Wood/cotton
  • Pk:
    2.000
Specifications
About this item
Wooden applicators with a double cotton bud.