ContactUsLinkComponent
Peristaltic pump tubing
Peristaltic pump tubing
N° de catalogue PERCB3140730
Fournisseur:  PerkinElmer
Peristaltic pump tubing
N° de catalogue PERCB3140730
Fournisseur:  PerkinElmer
Spécifications
  • Colour code:
    Red/red
  • Tubing int.Ø:
    1,14 mm (0,045")
  • Description:
    3 stopper
  • Pk:
    6
Spécifications
À propos de cet article
Pharmed tubing.
  • High fatigue strength under repeated reversed bending stresses
  • Non-toxic, biocompatible
  • Very low gas permeability
  • Well suited to acids and bases
Used for ICP-OES and ICP-MS applications.