Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

BOD standard solution

BOD standard solution, 210 mg/l, 1 litre bottle, 10
Catalog # WTWA252030
Supplier:   WTW
BOD standard solution
Catalog # WTWA252030
Supplier:  WTW
CAS Number:  
BOD standard solution
BOD standard solution

BOD standard solution, 210 mg/l, 1 litre bottle, 10

CAS Number:  
Price
177,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

 

Specifications

Specifications

  • Concentration:
    210 mg/l
  • Pack type:
    1 litre bottle
  • Pk:
    10
  • Description:
    BOD standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.