Orders LinkContactUsLinkComponent
Precision saccharometers
Precision saccharometers
Supplier:   Alla France
Precision saccharometers
Supplier:  Alla France
Certificates
Precision saccharometers
Precision saccharometers
5 Options Available Below
Product Details & Documents
Balling/ Brix precision saccharometers with red liquid filled thermometer.
Product Family Options
Product Information
  • Range
    Division
    Calibration temperature
    Length
    Availability
    Price
  • ALLA2900TC005/20QP
    Balling hydrometer
    Balling hydrometer
    –5 – +5 %
    0,1 %
    20 °C
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 154,00 €
    154,00 €
    1 * 1 ST
       154,00 €
  • ALLA2900TC000/20QP
    Balling hydrometer
    Balling hydrometer
    0 – 10 %
    0,1 %
    20 °C
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 154,00 €
    154,00 €
    1 * 1 ST
       154,00 €
  • ALLA2900TC010/20QP
    Balling hydrometer
    Balling hydrometer
    10 – 20 %
    0,1 %
    20 °C
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 154,00 €
    154,00 €
    1 * 1 ST
       154,00 €
  • ALLA2900TC020/20QP
    Balling hydrometer
    Balling hydrometer
    20 – 30 %
    0,1 %
    20 °C
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 154,00 €
    154,00 €
    1 * 1 ST
       154,00 €
  • ALLA2900TC030/20QP
    Balling hydrometer
    Balling hydrometer
    30 – 40 %
    0,1 %
    20 °C
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 154,00 €
    154,00 €
    1 * 1 ST
       154,00 €