Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Silver protein

Silver protein
Certificates

2 Options Available Below

Product Details & Documents

Protargol silver impregnation and neuronal selectivity for light and electron microscopic observations.

  • Assay: ~8% Ag basis
  • Titration: 7,0 to 9,0%
  • pH: 8,0 to 9,5 (10 mg/ml in water)
  • Solubility: 1 g in 10 ml of water

Silver proteinate (protargol) is used as a component of the Periodic Acid-ThioSemiCarbazide-Silver Proteinate (PA-TSC-SP) staining protocol for ultrastructural detection of carbohydrates such as glycogen, and proteoglycans.

Synonyms:  Silver colloidal, Silver nucleate, Silvol, Mild protargin, Colloidal Silver
  • CAS Number:9015-51-4
  • Formula:Ag (with protein)
  • MDL Number:MFCD00132268
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 76,60 €
    76,60 €
    1 * 5 g
       76,60 €

  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 238,00 €
    238,00 €
    1 * 25 g
       238,00 €