Blasticidin S hydrochloride ≥95%, crystalline solid
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:3513-03-9
:
Formula:C₁₇H₂₇ClN₈O₅
MDL Number:MFCD02091640
ADR:6.1,II
:458.904 g/mol
UN:2811
:GHS06
Storage Temperature:Freezer
Specification Test Results
- Purity:≥95%
- λmax:269 nm
Specification Test Results
Frequently Bought Together