Organic reference standard, bisphenol A D16, Dr Ehrenstorfer
Catalog # EHERC10655501
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:50 mg
- Standards:Reference standard
- CAS No.:96210-87-6
- Description:Bisphenol A D16
- Environmentally Preferable:
- Formula:C₁₅2H₁₆O₂
- Description:Bisphenol A D16
Specifications
About this item
Certifications: Produced in accordance to ISO 17034