Cellastim™ Human recombinant Albumin
BIRBORB134900-100
: Biorbyt
:
- Pk:100 g
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Storage conditions:–20 °C
- Protein synonyms:Albumin human
- Protein/peptide name:Albumin
- Purity:96%
- Molecular weight:67 kDa
Cellastim is a recombinant human serum albumin (HSA) that has been produced from an animal-free expression system.
- Replaces and outperforms FBS
- Well defined and consistent
- Animal-free, regulatory friendly
- Tested applications: ELISA
- Solubility (25 °C): cell culture tested
- Biological origin: expressed in rice
Cellastim has identical amino acid sequence and similar biophysical characteristics to the major HSA isoform found in human serum. Albumin is a cell culture supplement that functions as a carrier protein for fatty acids, growth factors, and trace minerals. Albumin also protects cells from damage due to toxins and by physical shear. Albumin is a common ingredient in many classical and custom media formulations. Albumin is used to reduce or eliminate serum and to support cell growth, health, and productivity.