

Tert-Butyle methyle oxyde ≥99.8% (GC, on anhydrous basis), PESTINORM® pour analyse des résidus de pesticides
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1634-04-4
:
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812
:88.15 g/mol
Melting Pt:–108,6 °C
Flash Pt:−28 °C
Storage Temperature:Température ambiante
UN:2398
ADR:3,II
:GHS02
:GHS07
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Appearance:Clear colorless liquid
- Assay (GC, on anhydrous basis):min. 99.8 %
- Identity (IR):Comply
- Residue after evaporation:max. 0.0005 % w/w
- Water (KF):max. 0.05 % w/w
- GC/ECD any Pesticide (as Lindane):max. 5 pg/ml
- Peroxides (as H₂0₂):max. 0.0005 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85932.290 spec_VWRC85932.320
Product specification
VWR® PESTINORM® Solvents
Product Literature
Frequently Bought Together