Chemical dispensing station, Chemstat 2.0
Catalog # 720-2942
Supplier: SERMO INDUSTRIALISATION
Specifications
- Description:Chemical dispensing station, Chemstat 2.0
- Pk:1
Specifications
About this item
Thanks to the new Chemstat 2,0, the search for safe and correct distribute and dilute of solvents is over.
- Extraction at the bottom for formaldehyde and at the top for solvents
- Archiving each action and user for traceability
- Ergonomic extendable carriers in the cabinet up to 60 liters
- Electric safety valve on the waste container
- Height-adjustable distribution faucets
Certifications: Anti-fire and explosive cabinet according to EN 14470-1 and TRGS 510.
Ordering information: Available with two or three dispensers that can all dilute.