ESD safety ankle boots, lace-up, Sander
Supplier: ELTEN
Certificates
About this item
100% metal- and leather-free, lightweight ESD safety ankle boot made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
- Climate-regulating, breathable textile lining
- Replaceable insole; orthopaedic insoles possible
- Full length fleece inlay sole (ESD Pro), black
- Metal-free puncture resistant midsole
- Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.
Specifications
- Catalog No:
- ELTN768321/44
- ELTN768321/45
- ELTN768321/43
- ELTN1768321/37
- ELTN768321/38
- ELTN768321/46
- ELTN1768321/35
- ELTN768321/47
- ELTN1768321/36
- ELTN768321/41
- ELTN768321/42
- ELTN768321/39
- ELTN768321/40
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001