VWR® Nitrile Green Nitrile gloves
Supplier: VWR Collection
Certificates
About this item
These single-use examination and disposable protective gloves are made from nitrile and are developed with sustainability in mind. They are suitable for use in medical applications, food processing and for general lab requirements in non hazardous areas.
- Free from chlorine, natural rubber latex proteins and rubber accelerators, particularly skin friendly for users with latex allergy
- Non sterile, powder-free
- Textured fingertips provide extra grip for wet or dry applications
- Synthetic inner coating for fast and easy glove donning
- Comfortable, strong glove material ensures stability and wearer comfort over long periods of time
Thanks to new sustainable and innovative production methods, you can reduce your eco-footprint while still wearing a glove that is skin friendly, and provides superior barrier protection against blood-borne pathogens and viruses. The production process has been redesigned to save gas and electricity, to save water (≥ 10% less), and reduce CO2 emissions (≥ 15% less). Made without the use of Chlorine gas during the entire production process.
Certifications: EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, MDR Class I; PPE Cat. III; AQL 1.5
Food contact approved.
Food contact approved.
Packaging: Supplied in a practical 180 or 200 piece big pack for simplified stock logistics, reduced replacement intervals in the workplace, and less packaging waste.
Caution: * Thickness is double-measured, minimum values
Specifications
- Catalog No:
- 112-5154
- 112-5156
- 112-5155
- 112-5158
- 112-5157
- Model:
- Nitrile PF Green
- Nitrile PF Green
- Nitrile PF Green
- Nitrile PF Green
- Nitrile PF Green
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Rolled
- Rolled
- Rolled
- Rolled
- Rolled
- Cuff thickness:
- 0,10 mm*
- 0,10 mm*
- 0,10 mm*
- 0,10 mm*
- 0,10 mm*
- Finger thickness:
- 0,14 mm*
- 0,14 mm*
- 0,14 mm*
- 0,14 mm*
- 0,14 mm*
- Palm thickness:
- 0,12 mm*
- 0,12 mm*
- 0,12 mm*
- 0,12 mm*
- 0,12 mm*
- EN norm:
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS
- Food contact approved:
- +
- +
- +
- +
- +
Specifications
Related Information
COC_VWRI112-5154_2022-10 L020263 2210 COC_VWRI112-5155_2023-08 L031147 2308 COC_VWRI112-5156_2021-07 L002232 2107 COC_VWRI112-5156_2101-05 L001245 2105
Food Contact Certificate for VWR Nitrile Green Gloves Recommendation on Chemical Resistance for VWR Nitrile Green Gloves Statement on Chemical Residues for VWR Nitrile Green Gloves VWR™ Nitrile Green Glove Sustainability Statement
Additional Product Details
VWR Collection Gloves VWR® Disposable Gloves VWR® Nitrile Green
Product Literature
Frequently Bought Together