Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, AVS TITRINORM volumetric solution
VWRS31950.370
:
- Pk:5 L
- Seq:5190
- Environmentally Preferable:
- NIST traceable:Confirmed
- Titer (20°C; real value 0.2 % accuracy):0.499 - 0.501 mol/l
: Potash caustic, Caustic potash
:
:56.11 g/mol
:1320
:215-181-3
:1814
:GHS05
:Ambient
:1310-58-3
:KOH
:MFCD00003553
:360
:1.023 g/cm³ (20 °C)
:8 II
:12,07806