Order Entry
Belgium
Orders LinkContactUsLinkComponent
SDS-MW gel buffer

SDS-MW gel buffer

SDS-MW gel buffer, proprietary formulation, 0,2% SDS, 8, 4
Catalog # BECLA30341
Marketsource
Supplier:   Beckman Coulter
SDS-MW gel buffer
Catalog # BECLA30341
Supplier:  Beckman Coulter
Marketsource
CAS Number:  
SDS-MW gel buffer

SDS-MW gel buffer, proprietary formulation, 0,2% SDS, 8, 4

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 4 ST
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    4
  • Description:
    SDS-MW gel buffer, proprietary formulation, 0,2% SDS
  • pH value:
    8

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.