Order Entry
Belgium
ContactUsLinkComponent
Lithium chloride ≥98%, TECHNICAL, purified
Irritant
Lithium chloride ≥98%, TECHNICAL, purified
Catalog # VWRS25009.361
Supplier:  VWR Collection
CAS Number:  
Irritant
Lithium chloride ≥98%, TECHNICAL, purified
Catalog # VWRS25009.361
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    6,5 kg
  • Seq:
    4126

Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

About this item

 :
 :42.39 g/mol
Merck Index:13,05550
Boiling Pt:1382 °C (1013 hPa)
Melting Pt:614 °C
Density:2.07 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-212-3
 :GHS07
CAS Number:7447-41-8
Formula:LiCl
MDL Number:MFCD00011078