Order Entry
Belgium
ContactUsLinkComponent
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Catalog # VWRS25631.320
Supplier:  VWR Collection
CAS Number:  
undefined
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Catalog # VWRS25631.320
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Acidity:
    Max. 0.0004 meq/g
  • Boiling point:
    39.0 - 40.5 °C
  • Density (20/4):
    1.322 - 1.327
  • Evaporation residue:
    Max. 20 ppm
  • Water:
    Max. 0.02 %

Specification Test Results

About this item

Synonyms: Methylene chloride
 :
 :84.93 g/mol
Melting Pt:–95 °C
EINECS:200-838-9
UN:1593
REACH:01-2119480404-41
 :GHS08
Storage Temperature:Ambient
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881
Boiling Pt:39.8 °C (1013 hPa)
Density:1.322 g/cm³ (20 °C)
 :0.1 % Stabilised with ethanol
ADR:6.1 III
Merck Index:12,06140