Orders LinkContactUsLinkComponent
Tablet counting triangles, SteriWare®
Tablet counting triangles, SteriWare®
Supplier:   Sampling Systems
Tablet counting triangles, SteriWare®
Supplier:  Sampling Systems
Certificates
Tablet counting triangles, SteriWare®
Tablet counting triangles, SteriWare®
2 Options Available Below
Product Details & Documents
The SteriWare® counting triangles are ideal for counting round tablets. They are manufactured and packed in a cleanroom and so can be used straight from the bag.
  • Made from FDA conforming PS
  • BSE/TSE free
  • Suitable for left handed and right handed operators
  • With an embossed scale to aid operation
Certifications: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
Packaging: Individually bagged in a heat-sealed laminate bag.
Product Family Options
Product Information
  • Colour
    W×D×H
    Availability
    Price
  • Sterile
    White
    230×230×13 mm
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 244,00 €
    244,00 €
    1 * 30 ST
       244,00 €
  • SAMP8066A-1
    Non-sterile
    Non-sterile
    White
    230×230×13 mm
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 183,00 €
    183,00 €
    1 * 30 ST
       183,00 €