Orders LinkContactUsLinkComponent
VWR®, Cell Holders
VWR®, Cell Holders
Supplier:   VWR Collection
VWR®, Cell Holders
Supplier:  VWR Collection
Certificates
VWR®, Cell Holders
VWR®, Cell Holders
28 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • For
    Availability
    Price
  • 634-6003
    Cell holder, 4-cell, for 10 mm square cuvette
    Cell holder, 4-cell, for 10 mm square cuvette
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
  • 634-6004
    Cell holder, 4-cell, for up to 50 mm square cuvette
    Cell holder, 4-cell, for up to 50 mm square cuvette
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 182,00 €
    182,00 €
    1 * 1 ST
       182,00 €
  • 634-1234
    Cell holder, 5-position auto cell changer, 10 - 100 mm automatic cell holder, five-position 10 - 100 mm cell holder (B) (for P9, M9)
    Cell holder, 5-position auto cell changer, 10 - 100 mm automatic cell holder, five-position 10 - 100 mm cell holder (B) (for P9, M9)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 804,00 €
    804,00 €
    1 * 1 ST
       804,00 €
  • 634-6005
    Cell holder, 4-cell, for up to 100 mm square cuvette
    Cell holder, 4-cell, for up to 100 mm square cuvette
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 515,00 €
    515,00 €
    1 * 1 ST
       515,00 €
  • 634-6006
    Cell holder, for cylindrical cell
    Cell holder, for cylindrical cell
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
  • 634-0778
    Cell holder (reference), 1 position for 10 mm standard cells
    Cell holder (reference), 1 position for 10 mm standard cells
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €
  • 634-6009
    Cell holder, test tube holder
    Cell holder, test tube holder
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 264,00 €
    264,00 €
    1 * 1 ST
       264,00 €
  • 634-0777
    Cell holder (sample), 1 position for 10 mm standard cells
    Cell holder (sample), 1 position for 10 mm standard cells
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €
  • 634-1135
    Cell holder, water-jacketed 1-position 10 mm cell holder, used for P4, PV4, M4, P6, M6
    Cell holder, water-jacketed 1-position 10 mm cell holder, used for P4, PV4, M4, P6, M6
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 364,00 €
    364,00 €
    1 * 1 ST
       364,00 €
  • 634-0687
    Cell holder, micro cell holder, 1 cell, beam height 15 mm
    Cell holder, micro cell holder, 1 cell, beam height 15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 515,00 €
    515,00 €
    1 * 1 ST
       515,00 €
  • 634-6051
    Cell holder, test tube holder (ø 8 - 22 mm) (B), for double beam only
    Cell holder, test tube holder (ø 8 - 22 mm) (B), for double beam only
    UV-6300PC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 259,00 €
    259,00 €
    1 * 1 ST
       259,00 €
  • 634-1133
    Cell holder, 1-position test tube holder, φ = 16 mm
    Cell holder, 1-position test tube holder, φ = 16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 229,00 €
    229,00 €
    1 * 1 ST
       229,00 €
  • 634-6049
    Cell holder, 10 mm water-jacketed one-cell holder (B), for double beam only
    Cell holder, 10 mm water-jacketed one-cell holder (B), for double beam only
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 489,00 €
    489,00 €
    1 * 1 ST
       489,00 €
  • 634-1232
    Auto cell holder, 8-position auto cell changer, 10 mm (for new P4, PV4, M4, P6, M6, P9, M9)
    Auto cell holder, 8-position auto cell changer, 10 mm (for new P4, PV4, M4, P6, M6, P9, M9)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 909,00 €
    909,00 €
    1 * 1 ST
       909,00 €
  • 634-6007
    Cell holder, 1 position, thermostattable, for 10 mm standard cells (external water bath required)
    Cell holder, 1 position, thermostattable, for 10 mm standard cells (external water bath required)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 348,00 €
    348,00 €
    1 * 1 ST
       348,00 €
  • 634-1233
    Auto cell holder, 5-position 10 - 100 mm cell holder (for VWR® P4, PV4, M4, P6, M6)
    Auto cell holder, 5-position 10 - 100 mm cell holder (for VWR® P4, PV4, M4, P6, M6)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 060,00 €
    1 060,00 €
    1 * 1 ST
       1 060,00 €
  • 634-6011
    Cell holder, solid sample holder (δ 1,5 - 4 mm; 1-position; for single beam and split beam beam only; sample size (L*W): 120*80 mm)
    Cell holder, solid sample holder (δ 1,5 - 4 mm; 1-position; for single beam and split beam beam only; sample size (L*W): 120*80 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 348,00 €
    348,00 €
    1 * 1 ST
       348,00 €
  • 634-6047
    Cell holder, one-cell holder for 100 mm square cuvette (B), for double beam only
    Cell holder, one-cell holder for 100 mm square cuvette (B), for double beam only
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 263,00 €
    263,00 €
    1 * 1 ST
       263,00 €
  • 634-6048
    Cell holder, cylindrical cell holder (ø 16 mm) (B), for double beam only
    Cell holder, cylindrical cell holder (ø 16 mm) (B), for double beam only
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 302,00 €
    302,00 €
    1 * 1 ST
       302,00 €
  • 634-1134
    Cell holder, water-jacketed 4-position 10 mm cell holder, used for P4, PV4, M4, P6, M6
    Cell holder, water-jacketed 4-position 10 mm cell holder, used for P4, PV4, M4, P6, M6
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 566,00 €
    566,00 €
    1 * 1 ST
       566,00 €
  • 634-1143
    Accessory sample room panel with port, used for P4, PV4, M4, P6, M6 (needed for 634-6007 and 634-6012)
    Accessory sample room panel with port, used for P4, PV4, M4, P6, M6 (needed for 634-6007 and 634-6012)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67,60 €
    67,60 €
    1 * 1 ST
       67,60 €
  • 634-6046
    Four-cell holder for up to 100 mm (10 - 100 mm) square cuvette (B) for double beam only
    Four-cell holder for up to 100 mm (10 - 100 mm) square cuvette (B) for double beam only
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 534,00 €
    534,00 €
    1 * 1 ST
       534,00 €
  • 634-6050
    Cell holder, 10 mm water-jacketed four-cell holder (B), for double beam only
    Cell holder, 10 mm water-jacketed four-cell holder (B), for double beam only
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 833,00 €
    833,00 €
    1 * 1 ST
       833,00 €
  • 634-6012
    Cell holder, 4 position, thermostattable, for 10 mm standard cells (external water bath required)
    Cell holder, 4 position, thermostattable, for 10 mm standard cells (external water bath required)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 515,00 €
    515,00 €
    1 * 1 ST
       515,00 €
  • 634-6045
    Cell holder, four-cell holder for 10 - 50 mm square cuvette (B), for double beam only
    Cell holder, four-cell holder for 10 - 50 mm square cuvette (B), for double beam only
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 277,00 €
    277,00 €
    1 * 1 ST
       277,00 €
  • 634-6010
    Cell holder, 8-postion, automatic for 10 mm standard cells
    Cell holder, 8-postion, automatic for 10 mm standard cells
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 937,00 €
    937,00 €
    1 * 1 ST
       937,00 €
  • 634-1136
    Cell holder, automatic 5-position, 10 mm
    Cell holder, automatic 5-position, 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 090,00 €
    1 090,00 €
    1 * 1 ST
       1 090,00 €
  • 634-6052
    Cell holder, solid sample holder (δ 1,5 - 4 mm, one-position) (B), for double beam only, sample size 120L×100W mm
    Cell holder, solid sample holder (δ 1,5 - 4 mm, one-position) (B), for double beam only, sample size 120L×100W mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 412,00 €
    412,00 €
    1 * 1 ST
       412,00 €