ContactUsLinkComponent
Suture, Dafilon (Aesculap®)
Suture, Dafilon (Aesculap®)
Catalog # AESCG118110
Supplier:  Aesculap
Suture, Dafilon (Aesculap®)
Catalog # AESCG118110
Supplier:  Aesculap
Specifications
  • Description:
    Dafilon black sutures
  • Length:
    15 cm
  • Pk:
    12
Specifications
About this item
Dafilon is a non-absorbable nylon monofilament suture.
  • Minimum tissue dragging
  • Inert material with good tissue compatibility
  • Excellent pliability
Suitable for skin closure in ophthalmic surgery, reconstructive surgery, and microsurgery.