Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Chromatography Vial Closures
PP cap for screw top autosampler vials, AQR™
PP cap for screw top autosampler vials, AQR™
PP cap for screw top autosampler vials, AQR™
Catalog # MICS9509S-10C-B
Supplier:
MICROSOLV
Expand image
PP cap for screw top autosampler vials, AQR™
Catalog # MICS9509S-10C-B
Supplier:
MICROSOLV
Specifications
For:
RSA™ or AQ™ screw top vials with 9-425 threads
Septum:
Non slit, flitted ultra pure silicone/PTFE septa
Cap colour:
Light blue
Pk:
100
Specifications
About this item
Product Details
Product Details