Orders LinkContactUsLinkComponent
pH buffer solutions, certified (DKD)
pH buffer solutions, certified (DKD)
pH buffer solutions, certified (DKD)
Fournisseur:  Mettler - Toledo
Certificates
pH buffer solutions, certified (DKD)
8 Options Available Below
Product Details & Documents
Certifications: The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the ideally suitable products for regulated industries. DKD is accepted by other accreditation bodies within the scope of the European Cooperation for Accreditation (EA) and the International Laboratory Accreditation Cooperation (ILAC).
Product Family Options
Product Information
  • pH value
    Pk Info
    Pk
    Disponibilité
    Prix
  • METT51350032
    DKD certified buffer solution
    DKD certified buffer solution
    4,01
    1× 250 ml
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 27,80 €
    27,80 €
    1 * 250 mL
       27,80 €
  • METT51350042
    DKD certified buffer solution
    DKD certified buffer solution
    4,01
    6× 250 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 101,00 €
    101,00 €
    1 * 6 ST
       101,00 €
  • METT51350034
    DKD certified buffer solution
    DKD certified buffer solution
    7,00
    1× 250 ml
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 27,80 €
    27,80 €
    1 * 250 mL
       27,80 €
  • METT51350044
    DKD certified buffer solution
    DKD certified buffer solution
    7,00
    6× 250 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 101,00 €
    101,00 €
    1 * 6 ST
       101,00 €
  • METT51350036
    DKD certified buffer solution
    DKD certified buffer solution
    9,21
    1× 250 ml
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 27,80 €
    27,80 €
    1 * 250 mL
       27,80 €
  • METT51350046
    DKD certified buffer solution
    DKD certified buffer solution
    9,21
    6× 250 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 101,00 €
    101,00 €
    1 * 6 ST
       101,00 €
  • METT51350038
    DKD certified buffer solution
    DKD certified buffer solution
    10,00
    1× 250 ml
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 27,80 €
    27,80 €
    1 * 250 mL
       27,80 €
  • METT51350048
    DKD certified buffer solution
    DKD certified buffer solution
    10,00
    6× 250 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 101,00 €
    101,00 €
    1 * 6 ST
       101,00 €