Orders LinkContactUsLinkComponent
Magnetic stirring bar, PTFE, triangular
Magnetic stirring bar, PTFE, triangular
Supplier:   COWIE
Magnetic stirring bar, PTFE, triangular
Supplier:  COWIE
Certificates
Magnetic stirring bar, PTFE, triangular
Magnetic stirring bar, PTFE, triangular
3 Options Available Below
Product Details & Documents
Magnetic stirring bar which is ideal for dissolving solids and mixing sediments due to the scraper-like action on the base of the container.
Documents
Product Family Options
Product Information
  • Length
    Ø
    Availability
    Price
  • 12 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 1,10 €
    1,10 €
    1 * 10 ST
       1,10 €
  • 25 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 16,80 €
    16,80 €
    1 * 5 ST
       16,80 €
  • 55 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 24,30 €
    24,30 €
    1 * 5 ST
       24,30 €